2-[4-Fluoro-3-(methylsulphonyl)phenyl]-1,3-dioxolane
Catalog # APOSPC510105-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:246.255 g/mol
Storage Temperature:Ambient
CAS Number:1354940-63-8
Formula:C₁₀H₁₁FO₄S
MDL Number:MFCD21332896