Methyl 4-(4-benzyl-1-piperazinyl)-3-fluorobenzoate
Catalog # APOSPC510082-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:328.381 g/mol
Storage Temperature:Ambient
CAS Number:1346597-56-5
Formula:C₁₉H₂₁FN₂O₂
MDL Number:MFCD20134208