1-[8-(4-Fluorophenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methanamine
Catalog # APOSPC510080-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:265.323 g/mol
Storage Temperature:Ambient
CAS Number:887979-07-9
Formula:C₁₅H₂₀FNO₂
MDL Number:MFCD06410588