1-(4-Fluorophenyl)-4-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1267968-10-4
:
Formula:C₁₃H₉FN₂O
MDL Number:MFCD18651705
:228.222 g/mol
Storage Temperature:Ambient
Frequently Bought Together