2-Hydroxy-4-methyl-6-(trifluoromethyl)nicotinamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:116548-07-3
:
Formula:C₈H₇F₃N₂O₂
MDL Number:MFCD16659045
:220.149 g/mol
Storage Temperature:Ambient
Frequently Bought Together