
8-Bromo-4-chloro-7-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2366994-26-3
:
Formula:C₁₀H₄BrClF₃N
MDL Number:MFCD31745749
:310.498 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together