
1-[4-(Trifluoromethyl)phenyl]imidazolidin-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:162748-21-2
:
Formula:C₁₀H₉F₃N₂O
MDL Number:MFCD22049301
:230.186 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together