
2-Chloro-3-fluoro-hydrazine hydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1138036-54-0
:
Formula:C₆H₆ClFN₂·ClH
MDL Number:MFCD18393091
:197.038 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together