

2-Bromo-5-(trifluoromethoxy)phenylisothiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2098276-18-5
:
Formula:C₈H₃BrF₃NOS
MDL Number:MFCD32200434
:298.08 g/mol
UN:2922
ADR:8+6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient
Frequently Bought Together