
3-(Bromodifluoromethyl)-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823188-41-5
:
Formula:C₈H₂BrClF₅N₃
MDL Number:MFCD29042499
:350.471 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together