2,6-Difluoro-4-(trifluoromethyl)phenyl 4-methoxyphenyl ether 95%
Catalog # APOSPC502837-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:304.212 g/mol
Storage Temperature:Ambient
CAS Number:2244084-78-2
Formula:C₁₄H₉F₅O₂
MDL Number:MFCD31629986