Order Entry
Czech Republic
ContactUsLinkComponent
4-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502831-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502831-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :308.631 g/mol
Storage Temperature:Ambient
CAS Number:2244087-56-5
Formula:C₁₃H₆ClF₅O
MDL Number:MFCD31629968