Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methoxyphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methoxyphenyl ether 95%
Catalog # APOSPC502822-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-methoxyphenyl ether 95%
Catalog # APOSPC502822-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :304.212 g/mol
Storage Temperature:Ambient
CAS Number:2244088-69-3
Formula:C₁₄H₉F₅O₂
MDL Number:MFCD31630090