Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-2,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-2,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502767-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2,6-dimethylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502767-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :381.135 g/mol
Storage Temperature:Ambient
CAS Number:2244084-69-1
Formula:C₁₅H₁₀BrF₅O
MDL Number:MFCD31630074