
2-Chloro-6-(trifluoromethoxy)benzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1261617-84-8
:
Formula:C₈H₄ClF₃O₃
MDL Number:MFCD18393799
:240.564 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together