
4-(Cyanomethyl)-3-fluorobenzene-1-sulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1551793-37-3
:
Formula:C₈H₅ClFNO₂S
MDL Number:MFCD26032776
ADR:8,III
:233.647 g/mol
UN:3261
:GHS05
Storage Temperature:Ambient
Frequently Bought Together