3-Nitro-4-(trifluoromethoxy)benzonitrile
Catalog # APOSPC501852-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.116 g/mol
Storage Temperature:Ambient
CAS Number:1214360-38-9
Formula:C₈H₃F₃N₂O₃
MDL Number:MFCD09745165