
4-Bromo-6-methoxy-2-(trifluoromethyl)benzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1980045-57-5
:
Formula:C₉H₆BrF₃N₂O
MDL Number:MFCD28348475
:295.056 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together