
3-Fluoro-5-(trifluoromethyl)benzene-1,2-diamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1217303-38-2
:
Formula:C₇H₆F₄N₂
MDL Number:MFCD28154972
:194.13 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together