
5-Bromo-6-chloro -2-(trifluoromethyl)-1H-benzimidazole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:39215-09-3
:
Formula:C₈H₃BrClF₃N₂
MDL Number:MFCD01658359
:299.475 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together