
4-(Bromomethyl)-3-(trifluoromethyl)benzoic acid, tech
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:859213-39-1
:
Formula:C₉H₆BrF₃O₂
MDL Number:MFCD22492213
:283.042 g/mol
Melting Pt:140…145 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together