
5-Bromo-2-fluoro-4-methylanisole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1780981-61-4
:
Formula:C₈H₈BrFO
MDL Number:MFCD28096600
:219.051 g/mol
Boiling Pt:54 °C (0.2 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together