
2,2,3,3-Tetrafluoro-1,4-benzodioxane-6-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:159276-63-8
:
Formula:C₉H₄F₄O₃
MDL Number:MFCD08275302
:236.12 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together