5-Bromo-2-fluoro-4-methoxybenzaldehyde
Catalog # APOSPC500445-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.034 g/mol
Storage Temperature:Ambient
CAS Number:473417-48-0
Formula:C₈H₆BrFO₂
MDL Number:MFCD11847135