
Methyl 3-hydroxy-5-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:796119-63-6
:
Formula:C₉H₇F₃O₃
MDL Number:MFCD12406849
:220.145 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together