
2-(Aminomethyl)-6-fluoroimidazo[1,2-a]pyridine
Catalog # APOSPC49658-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:165.168 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1020033-86-6
Formula:C₈H₈FN₃
MDL Number:MFCD09994905