Order Entry
Czech Republic
ContactUsLinkComponent
(1R)-1-(3,5-Difluorophenyl)ethylamine 97%
(1R)-1-(3,5-Difluorophenyl)ethylamine 97%
Catalog # APOSPC49497-1G
Supplier:  Apollo Scientific
CAS Number:  
(1R)-1-(3,5-Difluorophenyl)ethylamine 97%
Catalog # APOSPC49497-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :157.161 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
CAS Number:771465-40-8
Formula:C₈H₉F₂N
MDL Number:MFCD06761826