
2-Methyl-5-[4-(trifluoromethoxy)phenyl]-3-furoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:672930-56-2
:
Formula:C₁₃H₉F₃O₄
MDL Number:MFCD27937118
:286.203 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together