
3-Nitro-4-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:102844-90-6
:
Formula:C₈H₄F₃NO₃
MDL Number:MFCD05664203
:219.117 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together