
4-Bromo-2-iodo-1-(trifluoromethoxy)benzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:154065-33-5
:
Formula:C₇H₃BrF₃IO
MDL Number:MFCD18398076
:366.902 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together