

(2,4-Difluorophenyl)methanesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:179524-68-6
:
Formula:C₇H₅ClF₂O₂S
MDL Number:MFCD06798046
:226.628 g/mol
UN:3265
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together