

2-Fluoro-3-(trifluoromethyl)styrene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1298094-29-7
:
Formula:C₉H₆F₄
MDL Number:MFCD18909663
:190.138 g/mol
UN:1993
ADR:3,II
:GHS02
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together