

2-Hydroxy-2-(trifluoromethyl)butanoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:72114-82-0
:
Formula:C₅H₇F₃O₃
MDL Number:MFCD00190638
:172.102 g/mol
Boiling Pt:195…198 °C
Flash Pt:110 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together