Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-3-hydroxybenzeneboronic acid 96%
Irritant
2-Fluoro-3-hydroxybenzeneboronic acid 96%
Catalog # APOSPC48577-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Fluoro-3-hydroxybenzeneboronic acid 96%
Catalog # APOSPC48577-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :155.919 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:855230-60-3
Formula:C₆H₆BFO₃
MDL Number:MFCD06797222