(1R)-1-(3-Chloro-2-fluorophenyl)ethylamine hydrochloride 95+%
Catalog # APOSPC48549-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:210.076 g/mol
Storage Temperature:Ambient
CAS Number:1253792-97-0
Formula:C₈H₉ClFN·ClH
MDL Number:MFCD12758015