

(1S)-1-(3,5-Difluorophenyl)ethylamine hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1213128-98-3
:
Formula:C₈H₉F₂N·ClH
MDL Number:MFCD11101247
:193.622 g/mol
UN:3259
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together