(1R)-1-(3,5-Difluorophenyl)ethylamine hydrochloride 95+%
Catalog # APOSPC48536-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:193.622 g/mol
Storage Temperature:Ambient
CAS Number:321318-28-9
Formula:C₈H₉F₂N·ClH
MDL Number:MFCD11101200