
2-Fluoro-5-(trifluoromethoxy)nitrobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:124170-06-5
:
Formula:C₇H₃F₄NO₃
MDL Number:MFCD00042300
:225.097 g/mol
Flash Pt:>110 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together