
5-Chloro-4-hydroxy-6-(trifluoromethyl)pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:126538-83-8
:
Formula:C₅H₂ClF₃N₂O
MDL Number:MFCD09909761
:198.53 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together