
5-Bromo-2-methoxy-3-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1445995-86-7
:
Formula:C₉H₆BrF₃O₃
MDL Number:MFCD25541878
:299.041 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together