
Methyl 2-hydroxy-4-methyl-2-(trifluoromethyl)pent-4-enoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:117015-42-6
:
Formula:C₈H₁₁F₃O₃
MDL Number:MFCD08458103
:212.166 g/mol
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together