
(R)-3,3,4,4,5,5,5-Heptafluoropentan-2-ol 97%
Catalog # APOSPC450578-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:214.081 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:85571-83-1
Formula:C₅H₅F₇O
MDL Number:MFCD22569997