
1H,2H-1,2-Dichloroperfluorocyclopentane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1256353-27-1
:
Formula:C₅H₂Cl₂F₆
MDL Number:MFCD31560943
:246.966 g/mol
Boiling Pt:101 °C (760 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together