Order Entry
Czech Republic
ContactUsLinkComponent
N-Cyclohexyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide 95%
N-Cyclohexyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide 95%
Catalog # APOSPC450486-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Cyclohexyl-3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanamide 95%
Catalog # APOSPC450486-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :293.206 g/mol
Storage Temperature:Ambient
CAS Number:1980087-10-2
Formula:C₁₀H₁₃F₆NO₂
MDL Number:MFCD28901331