Order Entry
Czech Republic
ContactUsLinkComponent
 
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
Catalog # APOSPC450394-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,2H,2H-perfluorohexyl carbonate
Catalog # APOSPC450394-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :515.063 g/mol
Storage Temperature:Ambient
CAS Number:1980035-10-6
Formula:C₁₁H₈BrF₁₃O₃
MDL Number:MFCD28334466