
3-(2,2,2-Trifluoroethoxy)-5-(trifluoromethyl)benzoic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2375008-65-2
:
Formula:C₁₀H₆F₆O₃
MDL Number:MFCD32177069
:288.143 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together