Order Entry
Czech Republic
ContactUsLinkComponent
5-Nitro-4-(2,2,2-trifluoroacetamido)-2-(trifluoromethyl)benzoic acid
5-Nitro-4-(2,2,2-trifluoroacetamido)-2-(trifluoromethyl)benzoic acid
Catalog # APOSPC450256-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Nitro-4-(2,2,2-trifluoroacetamido)-2-(trifluoromethyl)benzoic acid
Catalog # APOSPC450256-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :346.14 g/mol
Storage Temperature:Ambient
CAS Number:1309602-22-9
Formula:C₁₀H₄F₆N₂O₅
MDL Number:MFCD19442153