3-(2,2-Difluoroethoxy)propionitrile
Catalog # APOSPC450254-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:135.112 g/mol
Storage Temperature:Ambient
CAS Number:1309602-19-4
Formula:C₅H₇F₂NO
MDL Number:MFCD19442116