
(R)-3,3,3-Trifluoropropane-1,2-diol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:148683-13-0
:
Formula:C₃H₅F₃O₂
MDL Number:MFCD16622004
Boiling Pt:87…88 °C (25 mm Hg)
Melting Pt:53…54 °C
:130.066 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together