
1,2-(1,1,2,2-Tetrafluoroethoxy)ethane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:358-39-4
:
Formula:C₆H₆F₈O₂
MDL Number:MFCD16619127
:262.098 g/mol
Boiling Pt:80 °C (50 mm Hg)
Density:1.48 g/cm³ (25 °C)
Flash Pt:66 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together